Learn More
Bochem™ 18/10 Stainless Steel Propeller Stirrers
Description
- For mixing of media with low and middle viscosity
- Ideal for homogenising and suspensioning
- Axial flow
Specifications
Specifications
| Diameter (Metric) | 50 mm |
| Diameter (Metric) Shaft | 8 mm |
| For Use With (Application) | Mixing of media |
| Length (Metric) | 400 mmH |
| Quantity | 1 Pack |
| Speed | Medium to High |
| Viscosity Rating | Low to Medium |