Learn More
Bochem™ 18/10 Stainless Steel Micro Spoon Spatulas, Scoop Shape
Product Code. 10758111
Change view
Click to view available options
Length (Metric):
150 mm
180 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
10758111 180 mm Each
10767911 150 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10758111 Supplier Bochem™ Supplier No. 3351
  • ONLINE EXCLUSIVE PRICE
    6.01 EUR
    4.44 EUR / Pack of 1
    Save 1.57 EUR
    26% Off
Estimated Shipment: 20-11-2026
Manufactured from 18/10 stainless steel. Bochem™ 18/10 Stainless Steel Micro Spoon Spatulas comes in two types.
  • 18/10 stainless steel
Specifications
Quantity 1 Ea.
Autoclavable Autoclavable
Shape Scoop
Product Type Micro Spoon Spatula
Length (Metric) Blade 40 mm
Width (Metric) 5 mm
Blade Material Stainless Steel (18/10)
Diameter (Metric) Outer 2mm
Dimensions (L x W) 180 x 5 mm
Material Stainless Steel (18/10)
Length (Metric) 180 mm