Learn More
Bochem™ 18/10 Stainless Steel Micro Spoon Spatulas
Product Code. 10269120
Change view
Click to view available options
Length (Metric):
150 mm
200 mm
Length (Metric) Blade:
5 mm (spoon) /45 mm (blade)
6 mm (spoon) / 45 mm (blade)
7 mm (spoon) / 40 mm (blade)
9 mm (spoon) / 40 mm (blade)
9 mm (spoon) / 45 mm (blade)
Width (Metric) Blade:
3 mm (spoon) / 5 mm (blade)
4 mm (spoon) / 5 mm (blade)
5 mm (spoon) / 5 mm (blade)
Unit Size:
Each
Pack of 10
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Length (Metric) Blade Width (Metric) Blade Unit Size
10269120 150 mm 7 mm (spoon) / 40 mm (blade) 5 mm (spoon) / 5 mm (blade) Each
10655801 150 mm 6 mm (spoon) / 45 mm (blade) 4 mm (spoon) / 5 mm (blade) Each
10383501 150 mm 5 mm (spoon) /45 mm (blade) 3 mm (spoon) / 5 mm (blade) Each
11815061 150 mm 5 mm (spoon) /45 mm (blade) 3 mm (spoon) / 5 mm (blade) Pack of 10
11825061 150 mm 9 mm (spoon) / 40 mm (blade) 5 mm (spoon) / 5 mm (blade) Pack of 10
10186310 150 mm 9 mm (spoon) / 40 mm (blade) 5 mm (spoon) / 5 mm (blade) Each
10373551 200 mm 9 mm (spoon) / 45 mm (blade) 5 mm (spoon) / 5 mm (blade) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 10269120 Supplier Bochem™ Supplier No. 3342
Estimated Shipment: 28-12-2026
Manufactured from 18/10 stainless steel. Bochem™ 18/10 Stainless Steel Micro Spoon Spatulas serve a wide variety of functions in the lab.
  • 18/10 Stainless steel
Spezifikation
Quantity 1 Ea.
Shape Spoon End, Blade End
Product Type Micro Spoon Spatula
Length (Metric) Blade 7 mm (spoon) / 40 mm (blade)
Width (Metric) 5 mm
Width (Metric) Blade 5 mm (spoon) / 5 mm (blade)
Dimensions (L x W) 150 x 5 mm
Material Stainless Steel (18/10)
Length (Metric) 150 mm