Learn More
Bochem™ 18/10 Stainless Steel Micro Spoon Spatulas, Spoon Shape
Product Code. 10557122
Change view
Click to view available options
Length (Metric):
130 mm
180 mm
185 mm
210 mm
Product Type:
Micro Spoon Spatula
Spoon Spatula
Diameter (Metric) Outer:
2mm
2.5mm
3mm
4mm
Unit Size:
Each
Pack of 10
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Product Type Diameter (Metric) Outer unitSize
10557122 210 mm Spoon Spatula 4mm Each
10420441 210 mm Micro Spoon Spatula 2mm Each
10770812 130 mm Micro Spoon Spatula 2mm Each
10094950 180 mm Micro Spoon Spatula 2.5mm Each
11835071 185 mm Micro Spoon Spatula 2mm Pack of 10
10721202 180 mm Spoon Spatula 3mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 10557122 Supplier Bochem™ Supplier No. 3214
In Stock
Add to basket
Edge
Add to basket
Manufactured from 18/10 stainless steel. Bochem™ 18/10 Stainless Steel Micro Spoon Spatulas comes in two types.
  •  Standard spatula
Specifications
Quantity 1 Ea.
Shape Spoon
Product Type Spoon Spatula
Diameter (Metric) Outer 4mm
Material Stainless Steel (18/10)
Length (Metric) 210 mm