Learn More
Bochem™ 18/10 Stainless Steel Chemical Spoon
Product Code. 10613341
Change view
Click to view available options
Length (Metric):
150 mm
180 mm
210 mm
250 mm
300 mm
Length (Metric) Blade:
32 mm
40 mm
45 mm
48 mm
Diameter (Metric) Outer:
1 mm
2 mm
5 mm
Unit Size:
Each
Pack of 10
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Length (Metric) Blade Diameter (Metric) Outer Unit Size
10613341 300 mm 48 mm 5 mm Each
10538881 150 mm 32 mm 5 mm Each
11865071 150 mm 32 mm 5 mm Pack of 10
10262091 150 mm 32 mm 1 mm Each
10485031 180 mm 32 mm 1 mm Each
10343981 210 mm 32 mm 1 mm Each
10373741 180 mm 40 mm 5 mm Each
11855071 210 mm 40 mm 5 mm Pack of 10
10169960 250 mm 45 mm 2 mm Each
10300681 250 mm 48 mm 5 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Product Code. 10613341 Supplier Bochem™ Supplier No. 3465
Sample Preparation and Storage Essentials
Use promo code 27996
View all promotions
Estimated Shipment: 30-10-2026
Add to basket
Add to basket
Stainless steel spoon for scooping chemicals.
  • Chemical spoon
  • With spatula handle
Specifications
Quantity 1 Ea.
Product Type Chemical Scoop, Double
Length (Metric) Blade 48 mm
Width (Metric) 35 mm
Diameter (Metric) Outer 5 mm
Material Stainless Steel (18/10)
Length (Metric) 300 mm