Learn More
Bochem™ 18/10 Stainless Steel Forceps, Sharp Tip
Product Code. 10788721
Change view
Click to view available options
Unit Size:
Each
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Unit Size
10788721 Each
10008051 Each
10252241 Each
10622591 Each
10373411 Each
10683341 Each
10050761 Each
10190301 Each
10212341 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 10788721 Supplier Bochem™ Supplier No. 1012
Sample Preparation and Storage Essentials
Use promo code 27996
View all promotions
Estimated Shipment: 09-10-2026
Add to basket
Add to basket
Assists with surgical procedures. Bochem™ 18/10 Stainless Steel Forceps, Sharp Tip are a very useful tool.
  • Stainless Steel
  • Various lengths
Specifications
Product Type Forceps
Material Stainless Steel (18/10)
Quantity 1 Pack
Tip Style Sharp
Length (Metric) 130 mm