Learn More
Bochem™ 18/10 E-Poli Crucible Tongs
Product Code. 10759461
Change view
Click to view available options
Length (Metric):
200 mm
300 mm
400 mm
500 mm
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
10759461 300 mm Each
10669662 200 mm Each
10160141 400 mm Each
10038710 500 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10759461 Supplier Bochem™ Supplier No. 2143
Estimated Shipment: 28-10-2026
Manufactured from stainless steel. Bochem™ 18/10 E-Poli Crucible Tongs are rugged.
  • Crucible tong
  • Rugged type
  • Stainless steel
Specifications
Product Type Crucible Tongs
Length (Metric) 300 mm
Material Stainless Steel (18/10)