Learn More
(16Beta)-21-(Acetyloxy)-17-hydroxy-16-methylpregna-1,4-diene-3,20-dione, TRC
Specifications
Specifications
| Chemical Name or Material | (16beta)-21-(Acetyloxy)-17-hydroxy-16-methylpregna-1,4-diene-3,20-dione |
| CAS | 24510-55-2 |
| Molecular Formula | C24H32O5 |
| Quantity | 5 mg |
| Synonym | 17,21-Dihydroxy-16beta-methyl-pregna-1,4-diene-3,20-dion, 21-Acetate (8CI) |
| InChI Formula | InChI=1S/C10H20N2O2/c1-10(2,3)14-9(13)12-8-4-6-11-7-5-8/h8,11H,4-7H2,1-3H3,(H,12,13) |
| SMILES | CC(C)(C)OC(=O)NC1CCNCC1 |
| IUPAC Name | tert-butyl N-piperidin-4-ylcarbamate |
| Molecular Weight (g/mol) | 400.51 |