missing translation for 'onlineSavingsMsg'
Learn More

Haloperidol, Thermo Scientific™

Brand:  Thermo Scientific Alfa Aesar J61688.14

Quantity 25g

Product Code. 15444239

  • 494.00 EUR / 25g

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

An antagonist for D2, D3, and D4 dopamine receptors that induces apoptosis of neurons in the striatum of rats.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

52-86-8
MFCD00051423
UN2811
haloperidol, haldol, aloperidin, serenace, aloperidol, brotopon, eukystol, einalon s, aloperidolo, galoperidol
OC1(CCN(CCCC(=O)C2=CC=C(F)C=C2)CC1)C1=CC=C(Cl)C=C1
375.87
CHEBI:5613
Haloperidol
C21H23ClFNO2
25g
14,4598
LNEPOXFFQSENCJ-UHFFFAOYSA-N
4-[4-(4-chlorophenyl)-4-hydroxypiperidin-1-yl]-1-(4-fluorophenyl)butan-1-one
3559
375.86
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.