Learn More
3M™ 1500 Graffiti Remover
Product Code. 17297610 Shop All 3M Products
Change view
Click to view available options
Volume (Metric):
500 mL
Unit Size:
Pack of 4
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
17297610 500 mL Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17297610 Supplier 3M™ Supplier No. 7000082039
  • ONLINE EXCLUSIVE PRICE
    315.00 EUR
    174.60 EUR / Pack of 4
    Save 140.40 EUR
    44% Off
 Restricted Item
Estimated Shipment: 23-12-2026
Removes graffiti, two types of remover depending on surface type
  • The solvent’s liquid form allows for quick penetration and rapid breakdown of graffiti
  • Does not contain NEP (N-Ethyl-pyrrolidone) or NMP (N-Methyl-2-pyrrolidone) used in other graffiti removers, which can pose a significant risk to health
  • Has little odour, enhancing user comfort and efficiency – unlike traditional chemical removers which emit powerful odours and harmful vapours
  • Low environmental impact as the product does not contain aromatic, aliphatic or chlorinated solvents
TRUSTED_SUSTAINABILITY
Specifications
Product Type Graffiti Remover
Volume (Metric) 500 mL