Learn More
(14bRS)-2-Methyl-1,2,3,4,10-14b-hexahydropyrazino[2,1-a]pyrido[2,3-c][2]benzazepine 2-Oxide, Mikromol™
Product Code. 30461265
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30461265 4 x 25 mg -
30052497 100 mg 100mg
30050924 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30461265 Supplier Mikromol Supplier No. MM1005.0200254AMP
 Restricted Item
Estimated Shipment: 30-11-2026
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(14bRS)-2-Methyl-1,2,3,4,10-14b-hexahydropyrazino[2,1-a]pyrido[2,3-c][2]benzazepine 2-Oxide, Mikromol
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Mirtazapine N-Oxide
CAS 155172-12-6
Analyte or Component Names Mirtazapine N-Oxide
Shipping Condition Room Temperature
Molecular Formula C17 H19 N3 O
Certifications/Compliance ISO 17025
Synonym Mirtazapine Imp. A (EP), Mirtazapine USP Related Compound B, Mirtazapine USP RC B, (14bRS)-2-Methyl-1,2,3,4,10-14b-hexahydropyrazino[2,1-a]pyrido[2,3-c][2]benzazepine 2-Oxide, Pyrazino[2,1-a]pyrido[2,3-c][2]benzazepine, 1,2,3,4,10,14b-hexahydro-2-methyl-, 2-oxide, 1,2,3,4,10,14b-Hexahydro-2-methylpyrazino[2,1-a]pyrido[2,3-c][2]benzazepine 2-oxide, Mirtazapine Impurity A
InChI Formula InChI=1S/C17H19N3O/c1-20(21)10-9-19-16(12-20)15-7-3-2-5-13(15)11-14-6-4-8-18-17(14)19/h2-8,16H,9-12H2,1H3
Solution Type Neat
SMILES C[N+]1([O-])CCN2C(C1)c3ccccc3Cc4cccnc24
IUPAC Name (14bRS)-2-Methyl-1,2,3,4,10-14b-hexahydropyrazino[2,1-a]pyrido[2,3-c][2]benzazepine 2-oxide
Molecular Weight (g/mol) 281.35
Quantity 4 x 25 mg
Formula Weight 281.1528 g/mol