Learn More
Fisherbrand™ Plastic Lock Bacti Caps
Product Code. 11547143 Shop All Fisherbrand Products
Change view
Click to view available options
Color:
Black
Blue
Red
White
Yellow
Unit Size:
Pack of 100
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
11547143 Blue Pack of 100
11527143 Red Pack of 100
11507143 Black Pack of 100
11517143 White Pack of 100
11537143 Yellow Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 11547143
Estimated Shipment: 21-09-2026
Add to basket
Add to basket
Made from chemically resistant virgin plastic, Bacti-Caps™ provide a firm, positive culture tube closure allowing a controlled gas interchange. They are ideal for use in science, research, and other laboratories.
  • Bacti-Caps™  for test tubes without edge
  • Provide a firm, positive culture tube closure allowing a controlled gas interchange, thus proving a more practical alternative to cotton wool plugs
  • Suitable for all standard test tube sizes, they fit rimless and plastic tubes with an outside diameter of 13mm, 16mm, 19mm, 25mm and 38mm 
  • The three flexible fins molded on the inner wall of each cap ensure a positive fit even when minor variations in tube size occur
  • Bacti-Caps™ are made from chemically resistant virgin plastic so they are unaffected by culture media, most acids, alkalis, alcohols and esters
  • Improved culture growth
  • Color coded
  • Controlled gas interchange
Specifications
Product Type Lock Bacti Cap
Autoclavable Autoclavable
Material Plastic
Color Blue
Quantity 100 Pack
Size (Metric) 13 mm
For Use With (Equipment) Test Tubes