Learn More
Labconco™ Drying Chamber for FreeZone™ Freeze-Dry Systems, PTFE-coated
Product Code. 15954650 Shop All Labconco Corporation Products
Change view
Click to view available options
No. of Ports:
12
16
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Ports unitSize
15954650 12 Each
15968132 16 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15954650 Supplier Labconco™ Supplier No. 7522801
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
Add to basket
Drying Chamber with corrosion-resistant interior for use on FreeZone™ Freeze Dryers.
  • Drying chamber made of type 304 stainless steel with corrosion-resistant PTFE-coated interior
  • Includes acrylic lid with neoprene gasket, latching pin, and neoprene valves with molded plastic knobs for 1/2 in. and 3/4 in. adapters
  • For use with FreeZone™ 1, 2.5, 4.5, 6, 8, 12, or 18 Liter Freeze Dryers
Specifications
Type Drying Chamber
No. of Shelves 0
Material PTFE-coated Stainless Steel
No. of Ports 12
For Use With (Application) To provide a means to attach flasks to freeze dry system or contain samples in bulk
For Use With (Equipment) FreeZone™ Freeze Dryers
Diameter (English) 8 in.
Height (English) 9.75 in.
Weight (English) 10 lb.