Learn More
Hanna Instruments™ Chemical Test Kit Replacement Reagents
Description
These high quality reagents are manufactured in Hanna's state-of-the-art facility and are clearly marked with the lot number and expiration date on each packet/bottle for traceability. Replacement titration reagent sets provide significant cost savings compared to purchasing an entirely new Test Kit.
Chemical Test Kit Replacement Reagents are available for the following Chemical Test Kits:
- Acidity Test Kit
- Alkalinity Test Kit
- Ascorbic Acid Test Kit
- Cardon Dioxide Test Kit
- Chloride Test Kit
- Dissolved Oxgen Test Kit
- Free and Total Chlorine (Low, Medium, High Range) Test Kit
- Free Chlorine Test Kit
- Total Chlorine Test Kit
- Iron (Medium Range) Test Kit
- Phosphate Test Kit
- Salinity Test Kit
- Sulfite Test Kit
- Total Hardness Test Kit
Specifications
Specifications
| Analytes | DPD (N,N-diethyl-p-phenylenediamine) |
| For Use With (Application) | Free Chlorine determination |
| For Use With (Equipment) | HI 3831F Free Chlorine Chemical Test Kit |
| Includes | Bottles with Protective Cap and Instructions |
| No. of Tests | 50 |
| Quantity | 1 Set |
| Range | 0 to 2.5mg/L (ppm) |
| Test Method | Titration |
| Type | Free Chlorine Test Kit |