missing translation for 'onlineSavingsMsg'
Learn More

11-Methyldodecanol, TRC

Product Code. 30482766
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30482766 1 g 1g
30464584 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30482766 Supplier Toronto Research Chemicals Supplier No. TRCM3037851G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85763-57-1
Molecular Formula C13H28O
Molecular Weight (g/mol) 200.36
Synonym 11-Methyl-1-dodecanol
SMILES CC(C)CCCCCCCCCCO

Specifications

Quantity 1 g
InChI Formula InChI=1S/C13H28O/c1-13(2)11-9-7-5-3-4-6-8-10-12-14/h13-14H,3-12H2,1-2H3
Chemical Name or Material 11-Methyldodecanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.