Learn More
11-Desethyl Irinotecan, TRC
Product Code. 30360326
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30360326 10 mg 10mg
30351456 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30360326 Supplier Toronto Research Chemicals Supplier No. TRCD28915510MG
 Restricted Item
Estimated Shipment: 08-10-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 103816-16-6
Molecular Formula C31 H34 N4 O6
Molecular Weight (g/mol) 558.62
Synonym [1,4'-Bipiperidine]-1'-carboxylic acid, (4S)-4-ethyl-3,4,12,14-tetrahydro-4-hydroxy-3,14-dioxo-1H-pyrano[3',4':6,7]indolizino[1,2-b]quinolin-9-yl ester, [1,4'-Bipiperidine]-1'-carboxylic acid, 4-ethyl-3,4,12,14-tetrahydro-4-hydroxy-3,14-dioxo-1H-pyrano[3',4':6,7]indolizino[1,2-b]quinolin-9-yl ester, (S)-, (4S)-4-Ethyl-3,4,12,14-tetrahydro-4-hydroxy-3,14-dioxo-1H-pyrano[3',4':6,7]indolizino[1,2-b]quinolin-9-yl [1,4'-bipiperidine]-1'-carboxylate, 1H-Pyrano[3',4':6,7]indolizino[1,2-b]quinoline, [1,4'-bipiperidine]-1'-carboxylic acid deriv., 10-[4-(1-Piperidino)-1-piperidinocarbonyloxy]camptothecin, (4S)-4-Ethyl-4-hydroxy-3,14-dioxo-3,4,12,14-tetrahydro-1H-pyrano[3',4':6,7]indolizino[1,2-b]quinolin-9-yl 1,4'-bipiperidine-1'-carboxylate, 11-Desethyl irinotecan
SMILES CC[C@@]1(O)C(=O)OCC2=C1C=C3N(Cc4cc5cc(OC(=O)N6CCC(CC6)N7CCCCC7)ccc5nc34)C2=O
Specifications
Quantity 10 mg
InChI Formula InChI=1S/C31H34N4O6/c1-2-31(39)24-16-26-27-20(17-35(26)28(36)23(24)18-40-29(31)37)14-19-15-22(6-7-25(19)32-27)41-30(38)34-12-8-21(9-13-34)33-10-4-3-5-11-33/h6-7,14-16,21,39H,2-5,8-13,17-18H2,1H3/t31-/m0/s1
Formula Weight 558.2478
Chemical Name or Material 11-Desethyl Irinotecan