missing translation for 'onlineSavingsMsg'
Learn More

11,17-Difuroate Mometasone FuroateMometasone Furoate Impurity E, TRC

Product Code. 30133543
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133543 100 mg 100mg
18070070 25 mg 25mg
18013150 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133543 Supplier Toronto Research Chemicals Supplier No. TRCD446190100MG
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1370190-33-2
Molecular Formula C32 H32 Cl2 O8
Molecular Weight (g/mol) 615.5
Synonym (11β,16α)-9,21-Dichloro-11,17-bis[(2-furanylcarbonyl)oxy]-16-methylpregna-1,4-diene-3,20-dione, 9,21-Dichloro-16α-methyl-3,20-dioxopregna-1,4-diene-11β,17-diyl bis(furan-2-carboxylate)
IUPAC Name [(8S,9R,10S,11S,13S,14S,16R,17R)-9-chloro-17-(2-chloroacetyl)-17-(furan-2-carbonyloxy)-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-11-yl] furan-2-carboxylate
SMILES C[C@@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(Cl)[C@H](C[C@]2(C)[C@@]1(OC(=O)c5occc5)C(=O)CCl)OC(=O)c6occc6

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C32H32Cl2O8/c1-18-14-22-21-9-8-19-15-20(35)10-11-29(19,2)31(21,34)26(41-27(37)23-6-4-12-39-23)16-30(22,3)32(18,25(36)17-33)42-28(38)24-7-5-13-40-24/h4-7,10-13,15,18,21-22,26H,8-9,14,16-17H2,1-3H3/t18-,21+,22+,26+,29+,30+,31+,32+/m1/s1
Formula Weight 614.1474
Chemical Name or Material 11,17-Difuroate Mometasone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.