Learn More
Tripentadecanoin-d5, TRC
Specifications
Specifications
| Chemical Name or Material | Tripentadecanoin-d5 |
| Molecular Formula | C48 D5 H87 O6 |
| Quantity | 10 mg |
| Synonym | Pentadecanoic Acid 1,2,3-Propanetriyl Ester-d5, 1,2,3-Tris(pentadecanoyl)glycerol-d5, Triglyceride-d5 PdPdPd, Pentadecanoic Acid 1,1',1''-(1,2,3-Propanetriyl) Ester-d5 |
| InChI Formula | InChI=1S/C48H92O6/c1-4-7-10-13-16-19-22-25-28-31-34-37-40-46(49)52-43-45(54-48(51)42-39-36-33-30-27-24-21-18-15-12-9-6-3)44-53-47(50)41-38-35-32-29-26-23-20-17-14-11-8-5-2/h45H,4-44H2,1-3H3/i43D2,44D2,45D |
| SMILES | [2H]C([2H])(OC(=O)CCCCCCCCCCCCCC)C([2H])(OC(=O)CCCCCCCCCCCCCC)C([2H])([2H])OC(=O)CCCCCCCCCCCCCC |
| IUPAC Name | [1,1,2,3,3-pentadeuterio-2,3-di(pentadecanoyloxy)propyl] pentadecanoate |
| Molecular Weight (g/mol) | 770.271 |
| Formula Weight | 769.721 |