Learn More
(10E)-10,11-Didehydro-11-deoxy-6-O-methylerythromycin, TRC
Product Code. 17940835
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17940835 25 mg 25mg
30134871 50 mg 50mg
17903905 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17940835 Supplier Toronto Research Chemicals Supplier No. TRCD29753025MG
 Restricted Item
Estimated Shipment: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 144604-03-5
Molecular Formula C38 H67 N O12
Molecular Weight (g/mol) 729.94
Synonym (10E)-10,11-Didehydro-11-deoxy-6-O-methylerythromycin A, Oxacyclotetradecane, erythromycin deriv.
IUPAC Name (3R,4S,5S,6R,7R,9R,11E,13S,14R)-6-[(2S,3R,4S,6R)-4-(dimethylamino)-3-hydroxy-6-methyloxan-2-yl]oxy-14-ethyl-13-hydroxy-4-[(2R,4R,5S,6S)-5-hydroxy-4-methoxy-4,6-dimethyloxan-2-yl]oxy-7-methoxy-3,5,7,9,11,13-hexamethyl-1-oxacyclotetradec-11-ene-2,10-dione
SMILES CC[C@H]1OC(=O)[C@H](C)[C@@H](O[C@H]2C[C@@](C)(OC)[C@@H](O)[C@H](C)O2)[C@H](C)[C@@H](O[C@@H]3O[C@H](C)C[C@@H]([C@H]3O)N(C)C)[C@@](C)(C[C@@H](C)C(=O)\C(=C\[C@]1(C)O)\C)OC
Specifications
Quantity 25 mg
InChI Formula InChI=1S/C38H67NO12/c1-15-27-36(8,44)17-20(2)29(40)21(3)18-38(10,46-14)33(51-35-30(41)26(39(11)12)16-22(4)47-35)23(5)31(24(6)34(43)49-27)50-28-19-37(9,45-13)32(42)25(7)48-28/h17,21-28,30-33,35,41-42,44H,15-16,18-19H2,1-14H3/b20-17+/t21-,22-,23+,24-,25+,26+,27-,28+,30-,31+,32+,33-,35+,36+,37-,38-/m1/s1
Formula Weight 729.4663
Chemical Name or Material (10E)-10,11-Didehydro-11-deoxy-6-O-methylerythromycin A