Learn More
Hellma™ Suprasil™ Quartz 104B Semi-Micro Cell Cuvettes
Product Code. 10495051
Change view
Click to view available options
Capacity (Metric):
1.4 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
10495051 1.4 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10495051 Supplier Hellma™ Supplier No. 104B1040
Only null left
Add to basket
Add to basket
Designed to hold samples for spectroscopic experiments and optical analysis. Hellma™ Suprasil™ Quartz 104B Semi-Micro Cell Cuvettes are supplied with a PTFE lid.
  • Sold in pairs
  • Very chemically resistant
  • Maintains shape, even at high temperatures up to approximately 1000°C
  • 80% transmission from 320 to 2500nm
Specifications
Type Semi-micro Cell
Capacity (Metric) 1.4 mL
Material Quartz Suprasil
Closure Material PTFE
Width (Metric) Interior 4mm
Quantity 1Each
Path Length 10 mm
Wavelength Range 200 to 2500 nm
Height (Metric) 45 mm
Width (Metric) 12.5 mm