missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fatty Acid Synthase/FASN Antibody (2A5), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26589-100ul
This item is not returnable.
View return policy
Description
Fatty Acid Synthase/FASN Monoclonal antibody specifically detects Fatty Acid Synthase/FASN in Human samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry
Specifications
| Fatty Acid Synthase/FASN | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, Immunocytochemistry | |
| 2A5 | |
| Flow Cytometry 1:20-1:200, ELISA, Immunocytochemistry/Immunofluorescence 1:20-1:200 | |
| EC 2.3.1, EC 2.3.1.85, EC 6.3.3.1, FASshort chain dehydrogenase/reductase family 27X, member 1, fatty acid synthase, MGC14367, MGC15706, OA-519, SDR27X1 | |
| A synthesized peptide derived from Human Fatty Acid Synthase/FASN [UniProt P49327] | |
| 100 μL | |
| Apoptosis, Cancer, Diabetes Research, Lipid and Metabolism, Neuroendocrinology | |
| 2194 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction