missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fatty Acid Synthase/FASN Antibody (2A5), Novus Biologicals™
Rabbit Monoclonal Antibody
343.00 EUR - 533.00 EUR
Specifications
| Antigen | Fatty Acid Synthase/FASN |
|---|---|
| Clone | 2A5 |
| Dilution | Flow Cytometry 1:20-1:200, ELISA, Immunocytochemistry/Immunofluorescence 1:20-1:200 |
| Applications | Flow Cytometry, ELISA, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30043827
|
Novus Biologicals
NBP3-26589-50ul |
50 μL |
343.00 EUR
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30043737
|
Novus Biologicals
NBP3-26589-100ul |
100 μL |
533.00 EUR
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Fatty Acid Synthase/FASN Monoclonal antibody specifically detects Fatty Acid Synthase/FASN in Human samples. It is validated for Flow Cytometry, ELISA, ImmunocytochemistrySpecifications
| Fatty Acid Synthase/FASN | |
| Flow Cytometry 1:20-1:200, ELISA, Immunocytochemistry/Immunofluorescence 1:20-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| EC 2.3.1, EC 2.3.1.85, EC 6.3.3.1, FASshort chain dehydrogenase/reductase family 27X, member 1, fatty acid synthase, MGC14367, MGC15706, OA-519, SDR27X1 | |
| A synthesized peptide derived from Human Fatty Acid Synthase/FASN [UniProt P49327] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 2A5 | |
| Flow Cytometry, ELISA, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Cancer, Diabetes Research, Lipid and Metabolism, Neuroendocrinology | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 2194 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title