Learn More
MPEG-2000-DSPE, TRC
Product Code. 17935055
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17935055 100 mg 100mg
17941245 250 mg 250mg
17932841 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17935055 Supplier Toronto Research Chemicals Supplier No. TRCM745845100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 147867-65-0
Molecular Formula (C2H4O)nC43H84NO10P
Molecular Weight (g/mol) 872.1
Synonym α-[6-Hydroxy-6-oxido-1,12-dioxo-9-[(1-oxooctadecyl)oxy]-5,7,11-trioxa-2-aza-6-phosphanonacos-1-yl]-ω-methoxy-Poly(oxy-1,2-ethanediyl)
IUPAC Name ethyl (E,4S)-2-methyl-4-[(2-methylpropan-2-yl)oxycarbonylamino]-5-(4-phenylphenyl)pent-2-enoate
SMILES CC(C)(C)OC(N[C@H](/C=C(C)/C(OCC)=O)CC(C=C1)=CC=C1C2=CC=CC=C2)=O
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C25H31NO4/c1-6-29-23(27)18(2)16-22(26-24(28)30-25(3,4)5)17-19-12-14-21(15-13-19)20-10-8-7-9-11-20/h7-16,22H,6,17H2,1-5H3,(H,26,28)/b18-16+/t22-/m1/s1
Formula Weight 409.2253
Chemical Name or Material MPEG-2000-DSPE