Learn More
Thermo Scientific™ 100% Graphite Ferrules for Thermo Scientific Instruments
Product Code. 10756144 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
Agilent Injectors and Non-MS Detectors
Thermo Scientific TRACE Ultra SSL Injectors and Non-MS Detectors
Diameter (Metric):
0.1 to 0.25 mm
0.32 mm
0.53 mm
3.175 mm
6.35 mm
Unit Size:
Pack of 10
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Diameter (Metric) Unit Size
10756144 Thermo Scientific TRACE Ultra SSL Injectors and Non-MS Detectors 0.1 to 0.25 mm Pack of 10
10444793 Thermo Scientific TRACE Ultra SSL Injectors and Non-MS Detectors 0.53 mm Pack of 10
10110943 Thermo Scientific TRACE Ultra SSL Injectors and Non-MS Detectors 0.32 mm Pack of 10
10409624 Agilent Injectors and Non-MS Detectors 6.35 mm Pack of 10
10311485 Agilent Injectors and Non-MS Detectors 3.175 mm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 10756144 Supplier Thermo Scientific™ Supplier No. 29053488
Save Up to 60% on Contract Testing Essentials
Use promo code 28025
View all promotions
Estimated Shipment: 28-09-2026
Add to basket
Edge
Add to basket
Ensure reliable connections for Thermo Scientific™ GC instruments with these Thermo Scientific™ 100% Graphite Ferrules for Thermo Scientific Instruments.
Ensure optimal performance of your Thermo Scientific™ GC instrument with Thermo Scientific™ Ferrules. They are available in three different materials and in various dimensions to accommodate a range of instruments and applications. All ferrules are are supplied in contaminant-free, individual blister packs.
Features
  • Available in 100% graphite, 15% graphite/85% vespel and SilTite metal
  • Ferrules for different types of injectors and detectors
TRUSTED_SUSTAINABILITY
Specifications
Type Ferrule
Description 0.1 to 0.25 mm I.D.; Thermo Instruments
Diameter (Metric) 0.1 to 0.25 mm
For Use With (Equipment) Thermo Scientific TRACE Ultra SSL Injectors and Non-MS Detectors
Material 100% Graphite
Quantity Pack of 10