Learn More
Hirschmann™ 100-150 mL Oxygen Bottle
Product Code. 11702293
Change view
Click to view available options
Volume (Metric):
100 to 150 mL
Unit Size:
Pack of 2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
11702293 100 to 150 mL Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11702293 Supplier Hirschmann™ Supplier No. 6500183
  • ONLINE EXCLUSIVE PRICE
    73.08 EUR
    67.90 EUR / Pack of 2
    Save 5.18 EUR
    7% Off
View all promotions
Estimated Shipment: 17-02-2027
With standard ground and chamfered glass stopper.
  • Acc. to Winkler
  • White graduation marks
  • Clear glass for excellent clarity
  • Precision adjusted
  • With label field
  • Individual serial number on the stopper and bottle
Specifications
Description Standard ground 14/23
Volume (Metric) 100 to 150 mL
Marking Spot Yes
Material Glass
Stopper Yes, glass
Height (Metric) 105 ± 10 mm