Learn More
10-Undecenoic Acid Ethyl Ester, TRC
Product Code. 30456614
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30456614 1 g 1g
30454411 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30456614 Supplier Toronto Research Chemicals Supplier No. TRCU7888251G
 Restricted Item
Estimated Shipment: 12-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 10-Undecenoic Acid Ethyl Ester
CAS 692-86-4
Molecular Formula C13H24O2
Quantity 1 g
Synonym Ethyl 10-Undecenoate, Ethyl Undecenoate
InChI Formula InChI=1S/C13H24O2/c1-3-5-6-7-8-9-10-11-12-13(14)15-4-2/h3H,1,4-12H2,2H3
SMILES CCOC(=O)CCCCCCCCC=C
IUPAC Name ethyl undec-10-enoate
Molecular Weight (g/mol) 212.33
Formula Weight 212.1776