Learn More
10-Hydroxydecanoic acid, 96%
Product Code. 15447238
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15447238 1 g 1g
15457238 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15447238 Supplier Thermo Scientific Alfa Aesar Supplier No. H60743.03
 Restricted Item
In Stock
Add to basket
CAS: 1679-53-4 | C10H20O3 | 188.27 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemische Identifikatoren
CAS 1679-53-4
Molecular Formula C10H20O3
Molecular Weight (g/mol) 188.27
MDL Number MFCD00010510
InChI Key YJCJVMMDTBEITC-UHFFFAOYSA-N
Synonym 10-hydroxydecanoate, decanoic acid, 10-hydroxy, unii-np03xo416b, 10-hydroxy capric acid, 10-hydroxy-decanoic acid, 1-hydroxydecanoicacid, 10-oh-capric acid, 10-hydroxycapric acid, 10-oh-decanoic acid, 10-hydroxydecanoicacid
PubChem CID 74300
ChEBI CHEBI:17409
IUPAC Name 10-hydroxydecanoic acid
SMILES OCCCCCCCCCC(O)=O
Spezifikation
Melting Point 75°C to 77°C
Linear Formula HO(CH2)9CO2H
Quantity 1 g
Formula Weight 188.26
Percent Purity 96%
Chemical Name or Material 10-Hydroxydecanoic acid