missing translation for 'onlineSavingsMsg'
Learn More

10-Hydroxybenzo[h]quinoline, 98%, Alfa Aesar™

Product Code. 11345485
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11345485 25 g 25g
11335485 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11345485 Supplier Thermo Scientific Alfa Aesar Supplier No. L19669.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Applications
10-Hydroxybenzo[h]quinolines is used to detect boronic acids in reactions and boron containing compounds on solid support. It is also used as a proton transfer-fluorescence probe to be applied to human serum albumin and beaver apomyoglobin.

Solubility
Soluble in acetone, chloroform and ethyl acetate.

Notes
Keep the container tightly sealed. Store in a cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33155-90-7
Molecular Formula C13H9NO
Molecular Weight (g/mol) 195.221
MDL Number MFCD00142350
InChI Key ZVFJWYZMQAEBMO-UHFFFAOYSA-N
Synonym 10-hydroxybenzo h quinoline, benzo h quinolin-10-ol, 10-hydroxy-benzo h quinoline, benzo h quinolin-10 1h-one, 4-hydroxy-5-azaphenanthrene, acmc-1cr1k, benzo h quinoline-10 1h-one, kesrrrlhhxxbrw-uhfffaoysa-n
PubChem CID 11816407
IUPAC Name 1H-benzo[h]quinolin-10-one
SMILES C1=CC(=O)C2=C3C(=CC=CN3)C=CC2=C1

Specifications

Melting Point 103°C to 105°C
Quantity 25 g
Beilstein 141770
Solubility Information Soluble in acetone,chloroform and ethyl acetate.
Formula Weight 195.22
Percent Purity 98%
Chemical Name or Material 10-Hydroxybenzo[h]quinoline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.