Learn More
10-Bromo-1-aminodecane, Hydrobromide, TRC
Specifications
Specifications
| Chemical Name or Material | 10-Bromo-1-aminodecane, Hydrobromide |
| CAS | 24566-82-3 |
| Molecular Formula | C10 H22 Br N . Br H |
| Quantity | 100 mg |
| InChI Formula | InChI=1S/C10H22BrN.BrH/c11-9-7-5-3-1-2-4-6-8-10-12;/h1-10,12H2;1H |
| SMILES | Br.NCCCCCCCCCCBr |
| IUPAC Name | 10-bromodecan-1-amine;hydrobromide |
| Molecular Weight (g/mol) | 317.1 |
| Formula Weight | 315.0197 |