missing translation for 'onlineSavingsMsg'
Learn More

1-Vinyl-2-pyrrolidone, Sigma-Aldrich™

Product Code. 15859640
Change view
Click to view available options
Quantity:
5 mL
250 mL
Unit Size:
250mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15859640 250 mL 250mL
15849640 5 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15859640 Supplier Sigma Aldrich Supplier No. 8085180250
 Restricted Item
Currently Unavailable: We have no information when this product will be available.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Stabilized with N,N′-di-sec-butyl-1,4-phenylenediamine for synthesis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88-12-0
Grade Research
Molecular Weight (g/mol) 111.14

Specifications

Melting Point 13°C to 14°C
Density 1.04 g/cm3 at 20°C
MDL Number MFCD00003197
Quantity 250 mL
Synonym 1-Vinyl-2-pyrrolidone, 1-Vinyl-2-pyrrolidinone, 1-Vinyl-2-pyrrolidone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.