missing translation for 'onlineSavingsMsg'
Learn More

1-Triethylsilyl-4-triethylsilyloxy-1-butyne, 97%

Product Code. 15420368
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15420368 25 g 25g
15410368 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15420368 Supplier Thermo Scientific Alfa Aesar Supplier No. H53488.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 160194-28-5 | C16H34OSi2 | 298.62 g/mol

1-Triethylsilyl-4-triethylsilyloxy-1-butyne is used as pharmaectical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Triethylsilyl-4-triethylsilyloxy-1-butyne is used as pharmaectical intermediates.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 160194-28-5
Molecular Formula C16H34OSi2
Molecular Weight (g/mol) 298.62
MDL Number MFCD00671376,MFCD09259947
InChI Key WAQSHASXNKNYSS-UHFFFAOYSA-N
Synonym 1-triethylsilyl-4-triethylsilyloxy-1-butyne, 1-triethylsilyl-4-triethlylsilyloxy-1-butyne, triethyl 4-triethylsilyl but-3-yn-1-yl oxy silane, triethyl 4-triethylsilyl but-3-inyloxy silane
PubChem CID 10935420
SMILES CC[Si](CC)(CC)OCCC#C[Si](CC)(CC)CC

Specifications

Quantity 25 g
Linear Formula (CH3CH2)3SiOCH2CH2C≡CSi(CH2CH3)3
IUPAC Name triethyl({[4-(triethylsilyl)but-3-yn-1-yl]oxy})silane
Formula Weight 298.62
Percent Purity 97%
Density 0.834
Chemical Name or Material 1-Triethylsilyl-4-triethylsilyloxy-1-butyne

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.