missing translation for 'onlineSavingsMsg'
Learn More

(1-Tetradecyl)triphenylphosphonium bromide, 97%

Product Code. 11376886
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11376886 100 g 100g
11366886 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11376886 Supplier Thermo Scientific Alfa Aesar Supplier No. L04311.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 25791-20-2 | C32H44BrP | 539.58 g/mol

(1-Tetradecyl)triphenylphosphonium bromide is used as an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(1-Tetradecyl)triphenylphosphonium bromide is used as an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

Solubility
Slightly soluble in Water.

Notes
Hygroscopic. Store away from oxidizing agents. Incompatible with moisture and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25791-20-2
Molecular Formula C32H44BrP
Molecular Weight (g/mol) 539.58
MDL Number MFCD00051857
InChI Key FUMBGFNGBMYHGH-UHFFFAOYSA-M
Synonym triphenyl tetradecyl phosphonium bromide, myristyltriphenylphosphonium bromide, 1-tetradecyl triphenylphosphonium bromide, triphenyl tetradecyl phosphanium bromide, tetradecyl triphenylphosphonium bromide, acmc-209gmk, tetradecyltriphenylphosphoniumbromide, triphenyl tetradecyl phosphoniumbromide, n-tetradecyltriphenylphosphonium bromide, phosphonium,triphenyltetradecyl-, bromide 1:1
PubChem CID 3084442
IUPAC Name triphenyl(tetradecyl)phosphanium;bromide
SMILES [Br-].CCCCCCCCCCCCCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 87°C to 89°C
Quantity 100 g
Beilstein 3582243
Sensitivity Hygroscopic
Solubility Information Slightly soluble in Water.
Formula Weight 539.59
Percent Purity 97%
Chemical Name or Material (1-Tetradecyl)triphenylphosphonium bromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.