CAS RN 25791-20-2
CAS RN 25791-20-2
IUPAC Name:
triphenyl(tetradecyl)phosphanium bromide
Synonyms:
triphenyl(tetradecyl)phosphanium bromide
Molecular Weight (g/mol):
539.58
Molecular Formula:
C32H44BrP
InChi Key:
FUMBGFNGBMYHGH-UHFFFAOYSA-M
SMILES:
[Br-].CCCCCCCCCCCCCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
(1-Tetradecyl)triphenylphosphonium bromide, 97%
CAS: 25791-20-2 Molecular Formula: C32H44BrP Molecular Weight (g/mol): 539.58 MDL Number: MFCD00051857 InChI Key: FUMBGFNGBMYHGH-UHFFFAOYSA-M Synonym: triphenyl tetradecyl phosphonium bromide,myristyltriphenylphosphonium bromide,1-tetradecyl triphenylphosphonium bromide,triphenyl tetradecyl phosphanium bromide,tetradecyl triphenylphosphonium bromide,acmc-209gmk,tetradecyltriphenylphosphoniumbromide,triphenyl tetradecyl phosphoniumbromide,n-tetradecyltriphenylphosphonium bromide,phosphonium,triphenyltetradecyl-, bromide 1:1 PubChem CID: 3084442 IUPAC Name: triphenyl(tetradecyl)phosphanium;bromide SMILES: [Br-].CCCCCCCCCCCCCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1