missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-1-Phenylethylamine, 98+%

Product Code. p-7022796
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
Product Code. Quantity unitSize
11407687 100 mL 100mL
11417687 500 mL 500mL
2 options
This item is not returnable. View return policy

Product Code. 11407687

Brand: Thermo Scientific Alfa Aesar A15711.AE

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 618-36-0 | C8H11N | 121.183 g/mol

Used for synthesis and as an emulsifying agent; used as a resolving agent and chiral intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used for synthesis and as an emulsifying agent; used as a resolving agent and chiral intermediate.

Solubility
Soluble in water at 20°C 4.2 g/L.

Notes
Air sensitive. Store away from air, acids and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Keep ignition sources away.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 618-36-0
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.183
MDL Number MFCD00008069
InChI Key RQEUFEKYXDPUSK-UHFFFAOYSA-N
Synonym 1-phenylethylamine, alpha-methylbenzylamine, alpha-phenylethylamine, dl-alpha-methylbenzylamine, 1-phenethylamine, alpha-aminoethylbenzene, 1-amino-1-phenylethane, 1-phenylethan-1-amine, a-methylbenzylamine, alpha-methylbenzenemethanamine
PubChem CID 7408
ChEBI CHEBI:670
IUPAC Name 1-phenylethanamine
SMILES CC(C1=CC=CC=C1)N

Specifications

Density 0.945
Boiling Point 69°C to 70°C (10 mmHg)
Flash Point 79°C (174°F)
Odor Odorless
Refractive Index 1.527
Quantity 100 mL
UN Number UN2735
Beilstein 636127
Sensitivity Air Sensitive
Merck Index 14,6026
Solubility Information Soluble in water at 20°C 4.2g/L.
Formula Weight 121.18
Percent Purity ≥98%
Chemical Name or Material (+/-)-1-Phenylethylamine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • (+/-)-1-Phenylethylamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : (+/-)-1-Phenylethylamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.