accessibility menu, dialog, popup

UserName

H3CNH2

CAS RN 618-36-0

IUPAC Name: 1-phenylethan-1-amine
Synonyms: (+/-)-α-methylbenzylamine 1-phenethylamine, (+-)- 1-phenethylamine, (r)- 1-phenethylamine, (s)- 1-fenylethylamin sumine 2079 1-phenethylamine, (R)- 1-phenethylamine, (S)- 1-phenethylamine 1-phenethylamine, (+-)-isomer
Molecular Weight (g/mol): 121.18
Molecular Formula: C8H11N
InChi Key: RQEUFEKYXDPUSK-UHFFFAOYNA-N
SMILES: CC(N)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (6)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (3)
  • (2)
Boiling Point 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (3)
  • (6)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)