Learn More
(+/-)-1-Phenylethyl isothiocyanate, 97%, Thermo Scientific™
Product Code. 11450461
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11450461 25 g 25g
11440461 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11450461 Supplier Thermo Scientific Alfa Aesar Supplier No. B20011.14
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 4478-92-6
Molecular Formula C9H9NS
Molecular Weight (g/mol) 163.24
MDL Number MFCD00022061
InChI Key QQCJPTVZIZVKEZ-UHFFFAOYNA-N
Synonym 1-phenylethyl isothiocyanate, 1-isothiocyanatoethyl benzene, alpha-methylbenzyl isothiocyanate, benzene, 1-isothiocyanatoethyl, dl-alpha-methylbenzyl isothiocyanate, dl-alpha-methylbenzylisothiocyanate, l-alpha-methylbenzyl isothiocyanate, +/--1-phenylethyl isothiocyanate, isothiocyanic acid, alpha-methylbenzyl ester, dl-.alpha.-methylbenzyl isothiocyanate
PubChem CID 20559
IUPAC Name 1-isothiocyanatoethylbenzene
SMILES CC(N=C=S)C1=CC=CC=C1
Specifications
Density 1.06
Boiling Point 132°C to 133°C (20 mmHg)
Flash Point 103°C (217°F)
Odor Pungent
Refractive Index 1.581
Quantity 25 g
UN Number UN2922
Beilstein 2207685
Sensitivity Moisture sensitive
Formula Weight 163.24
Percent Purity 97%
Chemical Name or Material (+/-)-1-Phenylethyl isothiocyanate