missing translation for 'onlineSavingsMsg'
Learn More

1-Phenyl-1-butanone, MP Biomedicals

Product Code. 11418385 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11418385 10 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11418385 Supplier MP Biomedicals Supplier No. 0522074605

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 495-40-9
Molecular Formula C10H12O
Quantity 10 g
Synonym butyrophenone, n-butyrophenone, 1-butanone, 1-phenyl, phenyl propyl ketone, 1-phenyl-1-butanone, propyl phenyl ketone, unii-186i11wb5b, 1-phenyl-butan-1-one, propylphenylketone
InChI Key FFSAXUULYPJSKH-UHFFFAOYSA-N
SMILES CCCC(=O)C1=CC=CC=C1
IUPAC Name 1-phenylbutan-1-one
Molecular Weight (g/mol) 148.205
PubChem CID 10315
Formula Weight 148.21
Chemical Name or Material 1-Phenyl-1-Butanon
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.