Learn More
1-Phenanthrol, TRC
Product Code. 30006677
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30006677 5 mg 5mg
30004631 25 mg 25mg
30006671 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30006677 Supplier Toronto Research Chemicals Supplier No. TRCP2950005MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2433-56-9
Molecular Formula C14 H10 O
Molecular Weight (g/mol) 194.23
Synonym 1-Phenanthrenol (9CI, ACI), 1-Phenanthrol (6CI, 7CI, 8CI), 1-Hydroxyphenanthrene, NSC 44471
IUPAC Name phenanthren-1-ol
SMILES Oc1cccc2c1ccc3ccccc23
Specifications
Quantity 5 mg
InChI Formula InChI=1S/C14H10O/c15-14-7-3-6-12-11-5-2-1-4-10(11)8-9-13(12)14/h1-9,15H
Formula Weight 194.0732
Chemical Name or Material 1-Hydroxyphenanthrene