missing translation for 'onlineSavingsMsg'
Learn More

1-Octene, Sigma-Aldrich™

Product Code. 15891261
Change view
Click to view available options
Quantity:
5 mL
250 mL
1 L
Unit Size:
1 L
250mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15891261 250 mL 250mL
15881261 5 mL 5mL
15801271 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15891261 Supplier Sigma Aldrich Supplier No. 8209280250
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For synthesis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-66-0
Grade Research
Molecular Formula CH3(CH2)5CH=CH2
Molecular Weight (g/mol) 112.21

Specifications

Melting Point -102°C
Density 0.72 g/cm3 at 20°C
MDL Number MFCD00009548
Quantity 250 mL
Synonym 1-Octene, 1-Octylene, Caprylene, 1-Octene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.