missing translation for 'onlineSavingsMsg'
Learn More

1-Octadecene, Sigma-Aldrich™

Product Code. 15871671
Change view
Click to view available options
Quantity:
100 mL
1 L
Unit Size:
1 L
100mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15871671 1 L 1 L
15861671 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15871671 Supplier Sigma Aldrich Supplier No. 8221121000
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For synthesis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-88-9
Grade Research
Molecular Formula CH3(CH2)15CH=CH2
Molecular Weight (g/mol) 252.48

Specifications

Melting Point 17°C to 18°C
Density 0.79 g/cm3 at 20°C
Boiling Point 315°C/1013 hPa
MDL Number MFCD00009003
Quantity 1 L
Synonym 1-Octadecene, 1-Octadecylene, 1-Octadecene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.