Learn More
1-Nitropyrene, TRC
Specifications
Specifications
| CAS | 5522-43-0 |
| Molecular Formula | C16 H9 N O2 |
| Quantity | 1 g |
| Synonym | Pyrene, 1-nitro-, 1-Nitropyrene, 3-Nitropyrene, NSC 81340 |
| InChI Formula | InChI=1S/C16H9NO2/c18-17(19)14-9-7-12-5-4-10-2-1-3-11-6-8-13(14)16(12)15(10)11/h1-9H |
| SMILES | [O-][N+](=O)c1ccc2ccc3cccc4ccc1c2c34 |
| IUPAC Name | 1-nitropyrene |
| Molecular Weight (g/mol) | 247.25 |
| Formula Weight | 247.0633 |
| Chemical Name or Material | 1-Nitropyrene |