missing translation for 'onlineSavingsMsg'
Learn More

1-Naphthyl Phosphate Monosodium Salt Monohydrate, MP Biomedicals™

Product Code. 11430625 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11430625 10 g 10g
11410625 1 g 1g
11420625 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11430625 Supplier MP Biomedicals Supplier No. 0219941010
  • ONLINE EXCLUSIVE PRICE
    365.00 EUR
    347.55 EUR / 10g
    Save 17.45 EUR
    4% Off
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Inhibits acid, alkaline and protein phosphatases. 1-Naphthyl Phosphate Monosodium Salt Monohydrate, MP Biomedicals is a non-specific phosphatase inhibitor. It is the preferred substrate for bioanalytical determinations of acid phosphatases, such as prosta

Chemical Identifiers

CAS 81012-89-7
Molecular Formula C10H7O4P
Molecular Weight (g/mol) 222.14
MDL Number MFCD00150615
InChI Key YNXICDMQCQPQEW-UHFFFAOYSA-L
Synonym 1-naphthyl phosphate monosodium salt monohydrate, sodium naphthalen-1-yl hydrogenphosphate hydrate, alpha-naphthyl acid phosphate monosodium, sodium hydrate naphthalen-1-yl hydrogen phosphate, 1-naphthyl phosphate sodium salt monohydrate, c10h8po4.na.h2o, sodium 1-naphthyl phosphate monohydrate, 1-naphthyl phosphate, monosodium salt monohydrate, naphthalen-1-yloxy sodiooxy phosphinic acid hydrate, 1-naphthyl phosphate monosodium salt monohydrate hplc
PubChem CID 45055387
SMILES [O-]P([O-])(=O)OC1=C2C=CC=CC2=CC=C1

Specifications

Physical Form Powder
Quantity 10 g
Solubility Information Soluble in water (50mg/mL)
IUPAC Name naphthalen-1-yl phosphate
Formula Weight 264.87g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.