Learn More
1-Naphthol, Sigma-Aldrich™
Product Code. 15841761
Change view
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15841761 1 kg 1kg
15831761 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15841761 Supplier Sigma Aldrich Supplier No. 8222891000
 Restricted Item
Estimated Shipment: 16-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 90-15-3
Grade Research
Molecular Weight (g/mol) 144.17
Specifications
Melting Point 94°C to 96°C
Density 1.28 g/cm3 at 20°C
Boiling Point 288°C/1013 hPa
MDL Number MFCD00003930
Quantity 1 kg
Synonym 1-Naphthol, 1-Hydroxynaphthalene, 1-Naphthol