missing translation for 'onlineSavingsMsg'
Learn More

1-Naphthol Beta-D-Glucuronide Sodium Salt, TRC

Product Code. 30008179
Change view
Click to view available options
Quantity:
1 mg
5 mg
25 mg
Unit Size:
1mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30008179 1 mg 1mg
30134050 25 mg 25mg
30003654 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30008179 Supplier Toronto Research Chemicals Supplier No. TRCN3680001MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 83833-12-9
Molecular Formula C16 H15 O7 . Na
Molecular Weight (g/mol) 342.28
Synonym 1-Naphthol-O-Beta-D-Glucuronide Sodium Salt
IUPAC Name sodium;(2S,3S,4S,5R,6S)-3,4,5-trihydroxy-6-naphthalen-1-yloxyoxane-2-carboxylate
SMILES [Na+].O[C@H]1[C@H](Oc2cccc3ccccc23)O[C@@H]([C@@H](O)[C@@H]1O)C(=O)[O-]

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C16H16O7.Na/c17-11-12(18)14(15(20)21)23-16(13(11)19)22-10-7-3-5-8-4-1-2-6-9(8)10;/h1-7,11-14,16-19H,(H,20,21);/q;+1/p-1/t11-,12-,13+,14-,16+;/m0./s1
Formula Weight 342.0715
Percent Purity >95%
Chemical Name or Material 1-Naphthyl-beta-D-Glucuronide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.