CAS RN 83833-12-9
CAS RN 83833-12-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1-Naphthol Beta-D-Glucuronide Sodium Salt, TRC
CAS: 83833-12-9 Molecular Formula: C16 H15 O7 . Na Molecular Weight (g/mol): 342.28 Synonym: 1-Naphthol-O-Beta-D-Glucuronide Sodium Salt IUPAC Name: sodium;(2S,3S,4S,5R,6S)-3,4,5-trihydroxy-6-naphthalen-1-yloxyoxane-2-carboxylate SMILES: [Na+].O[C@H]1[C@H](Oc2cccc3ccccc23)O[C@@H]([C@@H](O)[C@@H]1O)C(=O)[O-]