Learn More
1-(Naphthalen-1-yl)ethanol, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 1517-72-2 |
| Molecular Formula | C12H12O |
| MDL Number | MFCD00016806 |
| Quantity | 5 g |
| SMILES | CC(O)C1=C2C=CC=CC2=CC=C1 |
| Molecular Weight (g/mol) | 172.22 |
| Percent Purity | 98% |
| Chemical Name or Material | 1-(Naphthalen-1-yl)ethanol |