missing translation for 'onlineSavingsMsg'
Learn More

1-n-Hexyl-3-methylimidazolium tetrafluoroborate, 99%

Product Code. 11471840
Click to view available options
Quantity:
5 g
50 g
Unit Size:
50g
5g
This item is not returnable. View return policy

Product Code. 11471840

Brand: Thermo Scientific Alfa Aesar H27548.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 244193-50-8 | C10H19BF4N2 | 254.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 244193-50-8
Molecular Formula C10H19BF4N2
Molecular Weight (g/mol) 254.08
MDL Number MFCD03095459
InChI Key MFXLOVLEQJRXFP-UHFFFAOYSA-N
Synonym 1-hexyl-3-methylimidazolium tetrafluoroborate, 1-n-hexyl-3-methylimidazolium tetrafluoroborate, hmimbf4, c6mim bf4, acmc-1ckkl, hmim bf4, hmim+bf4, dsstox_cid_29108, dsstox_rid_83327, dsstox_gsid_49252
PubChem CID 2734179
IUPAC Name 1-hexyl-3-methylimidazol-3-ium;tetrafluoroborate
SMILES F[B-](F)(F)F.CCCCCCN1C=C[N+](C)=C1

Specifications

Melting Point -81°C
Density 1.149
Refractive Index 1.43
Quantity 50 g
Formula Weight 254.08
Percent Purity 99%
Chemical Name or Material 1-n-Hexyl-3-methylimidazolium tetrafluoroborate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.