missing translation for 'onlineSavingsMsg'
Learn More

1-n-Butyl-3-methylpyridinium tetrafluoroborate, 99%

Product Code. 11461610
Change view
Click to view available options
Quantity:
5 g
50 g
Unit Size:
50g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11461610 5 g 5g
11471610 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11461610 Supplier Thermo Scientific Alfa Aesar Supplier No. H26983.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 597581-48-1 | C10H16BF4N | 237.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 597581-48-1
Molecular Formula C10H16BF4N
Molecular Weight (g/mol) 237.05
MDL Number MFCD08458499
InChI Key UQEAMJVKASNOMA-UHFFFAOYSA-N
Synonym 1-butyl-3-methylpyridinium tetrafluoroborate, n-butyl-3-methylpyridinium tetrafluoroborate, 1-butyl-3-methylpyridin-1-ium tetrafluoroborate, acmc-20ak5s, dsstox_cid_29281, dsstox_rid_83400, dsstox_gsid_49325
PubChem CID 17841257
IUPAC Name 1-butyl-3-methylpyridin-1-ium;tetrafluoroborate
SMILES F[B-](F)(F)F.CCCC[N+]1=CC=CC(C)=C1

Specifications

Density 1.204
Quantity 5 g
Formula Weight 237.06
Percent Purity 99%
Chemical Name or Material 1-n-Butyl-3-methylpyridinium tetrafluoroborate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.