accessibility menu, dialog, popup

UserName

CAS RN 597581-48-1

BFFFFH3CNH3C

CAS RN 597581-48-1

IUPAC Name: 1-butyl-3-methylpyridin-1-ium; tetrafluoroboranuide
Synonyms: 1-butyl-3-methylpyridin-1-ium tetrafluoroborate
Molecular Weight (g/mol): 237.05
Molecular Formula: C10H16BF4N
InChi Key: UQEAMJVKASNOMA-UHFFFAOYSA-N
SMILES: F[B-](F)(F)F.CCCC[N+]1=CC=CC(C)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results