missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(Methylsulfonyl)imidazole, 98+%

Product Code. 11310094
Change view
Click to view available options
Quantity:
5g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11310094 5g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11310094 Supplier Alfa Aesar™ Supplier No. L00531.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 40736-26-3
Molecular Formula C4H6N2O2S
MDL Number MFCD00014496
Quantity 5g
Beilstein 4177637
Sensitivity Moisture sensitive
Synonym n-methanesulfonylimidazole, 1-methanesulfonylimidazole, 1-methylsulfonyl-1h-imidazole, 1-methylsulphonyl-1h-imidazole, 1-mesyl-1h-imidazole, acmc-20ak2l, 1-methylsulfonyl imidazole, 1h-imidazole,1-methylsulfonyl, 1-methylsulfonyl-1h-imidazole #
InChI Key ZBHRCOWHFHJZFP-UHFFFAOYSA-N
SMILES CS(=O)(=O)N1C=CN=C1
IUPAC Name 1-methylsulfonylimidazole
Molecular Weight (g/mol) 146.164
PubChem CID 521009
Formula Weight 146.17
Percent Purity ≥98%
Chemical Name or Material 1-(Methylsulfonyl)imidazole
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.